accessibility menu, dialog, popup

UserName

CAS RN 137-26-8

H3CNCH3SSSSNCH3CH3

CAS RN 137-26-8

IUPAC Name: N,N-dimethyl[(dimethylcarbamothioyl)disulfanyl]carbothioamide
Synonyms: tulisan hermal caswell no. 856 arasan-m arasan-M tetramethylthiocarbamoyldisulphide nocceler TT radothiram thioknock tetrapom
Molecular Weight (g/mol): 240.42
Molecular Formula: C6H12N2S4
InChi Key: KUAZQDVKQLNFPE-UHFFFAOYSA-N
SMILES: CN(C)C(=S)SSC(=S)N(C)C
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (2)