accessibility menu, dialog, popup

UserName

CAS RN 13670-99-0

H3COFF

CAS RN 13670-99-0

IUPAC Name: 1-(2,6-difluorophenyl)ethan-1-one
Synonyms: 1-(2,6-difluorophenyl)ethanone
Molecular Weight (g/mol): 156.13
Molecular Formula: C8H6F2O
InChi Key: VGIIILXIQLXVLC-UHFFFAOYSA-N
SMILES: CC(=O)C1=C(F)C=CC=C1F
Boiling Point 
  • (3)
  • (2)
Quantity 
  • (2)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (2)
  • (2)
  • (1)
  • (2)
  • (2)