accessibility menu, dialog, popup

UserName

FFFNSHN

CAS RN 136547-17-6

IUPAC Name: 6-(trifluoromethyl)-1,2-dihydropyrimidine-2-thione
Synonyms: 4-(trifluoromethyl)-3H-pyrimidine-2-thione
Molecular Weight (g/mol): 180.15
Molecular Formula: C5H3F3N2S
InChi Key: HDGJASCXBNRLGA-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=CC=NC(=S)N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results