accessibility menu, dialog, popup

UserName

CAS RN 13633-25-5

Br

CAS RN 13633-25-5

IUPAC Name: (4-bromobutyl)benzene
Synonyms: (4-bromobutyl)benzene
Molecular Weight (g/mol): 213.12
Molecular Formula: C10H13Br
InChi Key: XPBQQAHIVODAIC-UHFFFAOYSA-N
SMILES: BrCCCCC1=CC=CC=C1
Molecular Weight (g/mol) 
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)