accessibility menu, dialog, popup

UserName

CH3SCH3H3CN

CAS RN 13623-11-5

IUPAC Name: trimethyl-1,3-thiazole
Synonyms: trimethyl-1,3-thiazole
Molecular Weight (g/mol): 127.21
Molecular Formula: C6H9NS
InChi Key: BAMPVSWRQZNDQC-UHFFFAOYSA-N
SMILES: CC1=NC(C)=C(C)S1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results