accessibility menu, dialog, popup

UserName

CAS RN 136157-98-7

CH3CH3H3CCH2OOOHOCH3H2CH3CH3CH3COOOHCH3H3CCH3CH3CH3CH3OH3C

CAS RN 136157-98-7

IUPAC Name: 2-{[5,11,17,23-tetra-tert-butyl-27-(carboxymethoxy)-26,28-dimethoxypentacyclo[19.3.1.1³,⁷.1⁹,¹³.1¹⁵,¹⁹]octacosa-1(25),3,5,7(28),9(27),10,12,15(26),16,18,21,23-dodecaen-25-yl]oxy}acetic acid
Synonyms: {[5,11,17,23-tetra-tert-butyl-27-(carboxymethoxy)-26,28-dimethoxypentacyclo[19.3.1.1³,⁷.1⁹,¹³.1¹⁵,¹⁹]octacosa-1(25),3,5,7(28),9(27),10,12,15(26),16,18,21,23-dodecaen-25-yl]oxy}acetic acid
Molecular Weight (g/mol): 793.05
Molecular Formula: C50H64O8
InChi Key: GRSQCGICFHAMOA-UHFFFAOYSA-N
SMILES: COC1=C2CC3=CC(=CC(CC4=C(OC)C(CC5=C(OCC(O)=O)C(CC1=CC(=C2)C(C)(C)C)=CC(=C5)C(C)(C)C)=CC(=C4)C(C)(C)C)=C3OCC(O)=O)C(C)(C)C
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results