CAS RN 13587-26-3
CAS RN 13587-26-3
IUPAC Name:
copper(2+) diamine hexahydrate dihydrogen sulfate
Synonyms:
copper(2+) diamine hexahydrate dihydrogen sulfate
Molecular Weight (g/mol):
399.83
Molecular Formula:
CuH20N2O14S2
InChi Key:
IHEZHABINDDXAE-UHFFFAOYSA-L
SMILES:
N.N.O.O.O.O.O.O.[Cu++].OS([O-])(=O)=O.OS([O-])(=O)=O
1
–
1
of
1
results
1
Ammonium copper(II) sulfate hexahydrate, 99%
CAS: 13587-26-3 Molecular Formula: (NH4)2Cu(SO4)2·6H2O MDL Number: MFCD00150440 Synonym: Cupric ammonium sulfate