CAS RN 1357171-62-0
CAS RN 1357171-62-0
IUPAC Name:
N-({[1,1'-biphenyl]-4-yl}methyl)-6-phenyl-3-(pyridin-2-yl)-1,2,4-triazin-5-amine
Synonyms:
N-{[1,1'-biphenyl]-4-ylmethyl}-6-phenyl-3-(pyridin-2-yl)-1,2,4-triazin-5-amine
Molecular Weight (g/mol):
415.5
Molecular Formula:
C27H21N5
InChi Key:
QNRODODTMXCRKU-UHFFFAOYSA-N
SMILES:
C(NC1=C(N=NC(=N1)C1=CC=CC=N1)C1=CC=CC=C1)C1=CC=C(C=C1)C1=CC=CC=C1
1
–
3
of
3
results
1
N-([1,1'-Biphenyl]-4-ylmethyl)-6-phenyl-3-(pyridin-2-yl)-1,2,4-triazin-5-amine, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
N-([1,1'-Biphenyl]-4-ylmethyl)-6-phenyl-3-(pyridin-2-yl)-1,2,4-triazin-5-amine, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
N-([1,1'-Biphenyl]-4-ylmethyl)-6-phenyl-3-(pyridin-2-yl)-1,2,4-triazin-5-amine, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.