accessibility menu, dialog, popup

UserName

CAS RN 13530-50-2

OHPOOHOOHPOOHOOHPOOHOAl

CAS RN 13530-50-2

IUPAC Name: aluminium(3+) tridihydrogen phosphate
Synonyms: aluminium(3+) tridihydrogen phosphate
Molecular Weight (g/mol): 317.94
Molecular Formula: AlH6O12P3
InChi Key: RGPUVZXXZFNFBF-UHFFFAOYSA-K
SMILES: [Al+3].OP(O)([O-])=O.OP(O)([O-])=O.OP(O)([O-])=O
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results