CAS RN 13520-76-8
IUPAC Name:
tungsten dichloride dioxidandiide
Synonyms:
tungsten dichloride dioxide
Molecular Weight (g/mol):
286.74
Molecular Formula:
Cl2O2W
InChi Key:
DVBXMVWZPVBSJY-UHFFFAOYSA-L
SMILES:
[O--].[O--].[Cl-].[Cl-].[W]
1
–
1
of
1
results
1
Tungsten dichloride dioxide, 99%
CAS: 13520-76-8 Molecular Formula: WO2Cl2 MDL Number: MFCD00054136