accessibility menu, dialog, popup

UserName

CAS RN 135145-90-3

IUPAC Name: (2,5-dichlorophenyl)boronic acid
Synonyms: 2,5-dichlorophenylboronic acid
Molecular Weight (g/mol): 190.81
Molecular Formula: C6H5BCl2O2
InChi Key: NNTFPBXQPOQRBT-UHFFFAOYSA-N
SMILES: OB(O)C1=CC(Cl)=CC=C1Cl
Molecular Weight (g/mol) 
  • (3)
  • (5)
Quantity 
  • (3)
  • (2)
  • (3)
Percent Purity 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results