accessibility menu, dialog, popup

UserName

CAS RN 135-00-2

OS

CAS RN 135-00-2

IUPAC Name: phenyl(thiophen-2-yl)methanone
Synonyms: methanone, phenyl-2-thienyl-
Molecular Weight (g/mol): 188.24
Molecular Formula: C11H8OS
InChi Key: DWYFUJJWTRPARQ-UHFFFAOYSA-N
SMILES: O=C(C1=CC=CS1)C1=CC=CC=C1
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results