accessibility menu, dialog, popup

UserName

CAS RN 13472-81-6

BrNHOBr

CAS RN 13472-81-6

IUPAC Name: 3,5-dibromo-1,2-dihydropyridin-2-one
Synonyms: 3,5-dibromo-1H-pyridin-2-one
Molecular Weight (g/mol): 252.89
Molecular Formula: C5H3Br2NO
InChi Key: XIFRODWVHSZAMM-UHFFFAOYSA-N
SMILES: BrC1=CNC(=O)C(Br)=C1
Molecular Weight (g/mol) 
  • (1)
  • (4)
Percent Purity 
  • (1)
  • (3)
  • (1)
Quantity 
  • (2)
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (1)
  • (4)
  • (1)
  • (3)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)