accessibility menu, dialog, popup

UserName

CAS RN 13472-00-9

NH2NH2

CAS RN 13472-00-9

IUPAC Name: 4-(2-aminoethyl)aniline
Synonyms: 4-(2-aminoethyl)aniline
Molecular Weight (g/mol): 136.2
Molecular Formula: C8H12N2
InChi Key: LNPMZQXEPNWCMG-UHFFFAOYSA-N
SMILES: NCCC1=CC=C(N)C=C1
Molecular Weight (g/mol) 
  • (3)
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
  • (3)
Boiling Point 
  • (4)
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (3)
  • (3)
  • (2)
  • (2)
  • (2)
  • (2)
  • (3)
  • (4)
  • (2)
  • (2)
  • (2)