accessibility menu, dialog, popup

UserName

CH3NCONOO

CAS RN 13471-68-6

IUPAC Name: 2-isocyanato-1-methyl-4-nitrobenzene
Synonyms: 2-isocyanato-1-methyl-4-nitrobenzene
Molecular Weight (g/mol): 178.15
Molecular Formula: C8H6N2O3
InChi Key: PQFOUIMVTGBFRN-UHFFFAOYSA-N
SMILES: CC1=CC=C(C=C1N=C=O)[N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results