accessibility menu, dialog, popup

UserName

CAS RN 134620-00-1

IUPAC Name: palladium(2+) tetraamine dihydrogen carbonate
Synonyms: palladium(2+) tetraamine dibicarbonate
Molecular Weight (g/mol): 296.58
Molecular Formula: C2H14N4O6Pd
InChi Key: QYYPTZGPXKUKRM-UHFFFAOYSA-L
SMILES: N.N.N.N.[Pd++].OC([O-])=O.OC([O-])=O
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results