accessibility menu, dialog, popup

UserName

NOONBr

CAS RN 134604-07-2

IUPAC Name: 2-bromo-5-nitrobenzonitrile
Synonyms: 2-bromo-5-nitrobenzonitrile
Molecular Weight (g/mol): 227.02
Molecular Formula: C7H3BrN2O2
InChi Key: RKODNVITKISFKU-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC(C#N)=C(Br)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results