accessibility menu, dialog, popup

UserName

CAS RN 13455-21-5

H2OH2OFK

CAS RN 13455-21-5

IUPAC Name: potassium dihydrate fluoride
Synonyms: potassium dihydrate fluoride
Molecular Weight (g/mol): 94.13
Molecular Formula: FH4KO2
InChi Key: MZFXHSQFQLKKFD-UHFFFAOYSA-M
SMILES: O.O.[F-].[K+]
Percent Purity 
  • (3)
  • (3)
  • (3)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
Grade 
  • (2)
  • (3)
  • (3)
Physical Form 
  • (3)
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (5)
  • (3)
  • (3)
  • (3)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (3)
  • (3)
  • (3)
  • (3)
  • (2)