accessibility menu, dialog, popup

UserName

CAS RN 13454-71-2

CeOPOOO

CAS RN 13454-71-2

IUPAC Name: cerium(3+) phosphate
Synonyms: cerium(3+) phosphate
Molecular Weight (g/mol): 235.09
Molecular Formula: CeO4P
InChi Key: TYAVIWGEVOBWDZ-UHFFFAOYSA-K
SMILES: [Ce+3].[O-]P([O-])([O-])=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results