accessibility menu, dialog, popup

UserName

CAS RN 13451-11-1

BrBrBrBrBrTa

CAS RN 13451-11-1

IUPAC Name: tantalum(5+) pentabromide
Synonyms: tantalum(V) bromide
Molecular Weight (g/mol): 580.47
Molecular Formula: Br5Ta
InChi Key: GCPVYIPZZUPXPB-UHFFFAOYSA-I
SMILES: [Br-].[Br-].[Br-].[Br-].[Br-].[Ta+5]
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results