CAS RN 13392-24-0
CAS RN 13392-24-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
3
of
3
results
1
6-(L-1,2,3-Trihydroxybutyl)-pterin, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
6-(L-1,2,3-Trihydroxybutyl)-pterin, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
6-(L-1,2,3 - Trihydroxybutyl) -pterin, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.