CAS RN 133909-97-4
CAS RN 133909-97-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
N-Triphenylmethyl-5-[4'-methylbiphenyl-2-yl]tetrazole, TRC
CAS: 133909-97-4 Molecular Formula: C33 H26 N4 Molecular Weight (g/mol): 478.59 Synonym: 2H-Tetrazole, 5-(4'-methyl[1,1'-biphenyl]-2-yl)-2-(triphenylmethyl)-,5-(4'-Methyl[1,1'-biphenyl]-2-yl)-2-(triphenylmethyl)-2H-tetrazole IUPAC Name: 5-[2-(4-methylphenyl)phenyl]-2-trityltetrazole SMILES: Cc1ccc(cc1)c2ccccc2c3nnn(n3)C(c4ccccc4)(c5ccccc5)c6ccccc6