accessibility menu, dialog, popup

UserName

CAS RN 133550-34-2

IUPAC Name: 2-cyano-3-(3,4-dihydroxyphenyl)-N-(3-phenylpropyl)prop-2-enamide
Synonyms: 2-cyano-3-(3,4-dihydroxyphenyl)-N-(3-phenylpropyl)prop-2-enamide
Molecular Weight (g/mol): 322.36
Molecular Formula: C19H18N2O3
InChi Key: GSQOBTOAOGXIFL-UHFFFAOYSA-N
SMILES: OC1=C(O)C=C(C=C(C#N)C(=O)NCCCC2=CC=CC=C2)C=C1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results