accessibility menu, dialog, popup

UserName

H3COOSCH3

CAS RN 13327-56-5

IUPAC Name: ethyl 3-(methylsulfanyl)propanoate
Synonyms: ethyl 3-(methylsulfanyl)propanoate
Molecular Weight (g/mol): 148.22
Molecular Formula: C6H12O2S
InChi Key: YSNWHRKJEKWJNY-UHFFFAOYSA-N
SMILES: CCOC(=O)CCSC
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results