CAS RN 132907-72-3
CAS RN 132907-72-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Ramosetron Hydrochloride, TRC
CAS: 132907-72-3 Molecular Formula: C17 H17 N3 O . Cl H Molecular Weight (g/mol): 315.8 Synonym: (1-Methyl-1H-indol-3-yl)[(6R)-4,5,6,7-tetrahydro-1H-benzimidazol-6-yl]methanone Hydrochloride,(-)-(R)-5-[(1-Methyl-1H-indol-3-yl)carbonyl]-4,5,6,7-tetrahydro-1H- benzimidazole Monohydrochloride,(R)-(-)-Ramosetron Hydrochloride,Irribow,YM 060; IUPAC Name: (1-methylindol-3-yl)-[(5R)-4,5,6,7-tetrahydro-1H-benzimidazol-5-yl]methanone;hydrochloride SMILES: Cl.Cn1cc(C(=O)[C@@H]2CCc3[nH]cnc3C2)c4ccccc14