accessibility menu, dialog, popup

UserName

CAS RN 131748-14-6

NClFFF

CAS RN 131748-14-6

IUPAC Name: 4-chloro-2-(trifluoromethyl)pyridine
Synonyms: 4-chloro-2-(trifluoromethyl)pyridine
Molecular Weight (g/mol): 181.54
Molecular Formula: C6H3ClF3N
InChi Key: FZINIBTZNSPWQR-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=NC=CC(Cl)=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results