CAS RN 1314-11-0
CAS RN 1314-11-0
IUPAC Name:
strontium(2+) oxidandiide
Synonyms:
strontium oxide
Molecular Weight (g/mol):
103.62
Molecular Formula:
OSr
InChi Key:
IATRAKWUXMZMIY-UHFFFAOYSA-N
SMILES:
[O--].[Sr++]
1
–
2
of
2
results
1
Strontium oxide, tech.
CAS: 1314-11-0 Molecular Formula: OSr Molecular Weight (g/mol): 103.619 MDL Number: MFCD00049558 InChI Key: UFQXGXDIJMBKTC-UHFFFAOYSA-N PubChem CID: 518709 IUPAC Name: oxostrontium SMILES: O=[Sr]
2
Strontium oxide, 99.5% (metals basis), SrO ^=97%
CAS: 1314-11-0 Molecular Formula: OSr Molecular Weight (g/mol): 103.619 MDL Number: MFCD00049558 InChI Key: UFQXGXDIJMBKTC-UHFFFAOYSA-N PubChem CID: 518709 IUPAC Name: oxostrontium SMILES: O=[Sr]