CAS RN 1312092-82-2
CAS RN 1312092-82-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
2-[(4,6-Dimethyl-2-pyrimidinyl)oxy]-3,3-diphenyl-2-propenoic Acid, TRC
CAS: 1312092-82-2 Molecular Formula: C21H18N2O3 Molecular Weight (g/mol): 346.38 Synonym: 2-((4,6-Dimethylpyrimidin-2-yl)oxy)-3,3-diphenylacrylic Acid,Ambrisentan Impurity E SMILES: Cc1cc(C)nc(OC(=C(c2ccccc2)c3ccccc3)C(=O)O)n1