CAS RN 13080-90-5
IUPAC Name:
bicyclo[2.2.1]hept-5-en-2-ol
Synonyms:
5-norbornen-2-ol (exo)
Molecular Weight (g/mol):
110.16
Molecular Formula:
C7H10O
InChi Key:
MKOSBHNWXFSHSW-UHFFFAOYNA-N
SMILES:
OC1CC2CC1C=C2
1
–
1
of
1
results
1
5-Norbornen-2-ol, mixture of endo and exo, 98+%
CAS: 13080-90-5 Molecular Formula: C7H10O Molecular Weight (g/mol): 110.156 MDL Number: MFCD00167566 InChI Key: MKOSBHNWXFSHSW-UHFFFAOYSA-N Synonym: 5-norbornene-2-ol,bicyclo 2.2.1 hept-5-en-2-ol,5-norbornen-2-ol,5-norbornen-2-ol, exo,exo-2-norbornenol,bicyclo 2.2.1 hept-2-en-5-ol,bicyclo 2.2.1 hept-5-en-2-ol, exo,5-norbornen-2-ol exo PubChem CID: 96066 IUPAC Name: bicyclo[2.2.1]hept-2-en-5-ol SMILES: C1C2CC(C1C=C2)O