accessibility menu, dialog, popup

UserName

CAS RN 129865-52-7

OSNN

CAS RN 129865-52-7

IUPAC Name: 2-(piperidin-1-yl)-1,3-thiazole-5-carbaldehyde
Synonyms: 2-(piperidin-1-yl)-1,3-thiazole-5-carbaldehyde
Molecular Weight (g/mol): 196.27
Molecular Formula: C9H12N2OS
InChi Key: NWOAHSDBLWFXAG-UHFFFAOYSA-N
SMILES: O=CC1=CN=C(S1)N1CCCCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results