CAS RN 127657-97-0
IUPAC Name:
[(2,2-dimethoxyethoxy)methyl]benzene
Synonyms:
[(2,2-dimethoxyethoxy)methyl]benzene
Molecular Weight (g/mol):
196.25
Molecular Formula:
C11H16O3
InChi Key:
QITBBLPNYKECAZ-UHFFFAOYSA-N
SMILES:
COC(COCC1=CC=CC=C1)OC
1
–
1
of
1
results
1
Benzyloxyacetaldehyde dimethyl acetal, 97%
CAS: 127657-97-0 Molecular Formula: C11H16O3 Molecular Weight (g/mol): 196.246 MDL Number: MFCD00082233 InChI Key: QITBBLPNYKECAZ-UHFFFAOYSA-N Synonym: benzyloxyacetaldehyde dimethyl acetal,2,2-dimethoxyethoxy methyl benzene,benzyloxyacetaldehyde dimethylacetal,benzene, 2,2-dimethoxyethoxy methyl,acmc-20alp4,2,2-dimethoxyethylbenzyl ether,2-benzyloxy-1,1-dimethoxyethane;,benzyloxy-acetaldehyd-dimethylacetal; PubChem CID: 7021458 IUPAC Name: 2,2-dimethoxyethoxymethylbenzene SMILES: COC(COCC1=CC=CC=C1)OC