CAS RN 1271-24-5
IUPAC Name:
λ²-chromium(2+) bis(cyclopenta-2,4-dien-1-ide)
Synonyms:
chromocene
Molecular Weight (g/mol):
182.19
Molecular Formula:
C10H10Cr
InChi Key:
TYYBBNOTQFVVKN-UHFFFAOYSA-N
SMILES:
[Cr++].[CH-]1C=CC=C1.[CH-]1C=CC=C1
1
–
1
of
1
results
1
Bis(cyclopentadienyl)chromium, sublimed, 97+%
CAS: 1271-24-5 Molecular Formula: C10H10Cr Molecular Weight (g/mol): 182.19 MDL Number: MFCD00013750 InChI Key: TYYBBNOTQFVVKN-UHFFFAOYSA-N Synonym: Chromocene IUPAC Name: Bis(cyclopentadienyl)chromium SMILES: [Cr++].[CH-]1C=CC=C1.[CH-]1C=CC=C1