accessibility menu, dialog, popup

UserName

CH3CH3SOONHOHO

CAS RN 126146-01-8

IUPAC Name: 4-(2,5-dimethylbenzenesulfonamido)benzoic acid
Synonyms: 4-(2,5-dimethylbenzenesulfonamido)benzoic acid
Molecular Weight (g/mol): 305.35
Molecular Formula: C15H15NO4S
InChi Key: MGFQTAXIYWVXQN-UHFFFAOYSA-N
SMILES: CC1=CC(=C(C)C=C1)S(=O)(=O)NC1=CC=C(C=C1)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results