CAS RN 1261393-19-4
CAS RN 1261393-19-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Candesartan Cilexetil-d11, TRC
CAS: 1261393-19-4 Molecular Formula: C33H23D11N6O6 Molecular Weight (g/mol): 621.73 Synonym: 2-Ethoxy-1-[[2'-(1H-tetrazol-5-yl)[1,1'-biphenyl]-4-yl]methyl]-1H-benzimidazole-7-carboxylic Acid 1-[[(Cyclohexyl-d11-oxy)carbonyl]oxy]ethyl Ester,Atacand-d11,Blopress-d11,TCV 116-d11; IUPAC Name: 1-(1,2,2,3,3,4,4,5,5,6,6-undecadeuteriocyclohexyl)oxycarbonyloxyethyl 2-ethoxy-3-[[4-[2-(2H-tetrazol-5-yl)phenyl]phenyl]methyl]benzimidazole-4-carboxylate SMILES: O=C(OC(C)OC(OC1([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C1([2H])[2H])=O)C2=CC=CC3=C2N(CC4=CC=C(C5=CC=CC=C5C6=NN=NN6)C=C4)C(OCC)=N3