accessibility menu, dialog, popup

UserName

CAS RN 125650-44-4

IUPAC Name: 5-cyclobutyl-5-phenylimidazolidine-2,4-dione
Synonyms: 5-cyclobutyl-5-phenylimidazolidine-2,4-dione
Molecular Weight (g/mol): 230.27
Molecular Formula: C13H14N2O2
InChi Key: YFMSPNSDLBGLRF-UHFFFAOYNA-N
SMILES: O=C1NC(=O)C(N1)(C1CCC1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results