CAS RN 1242258-49-6
CAS RN 1242258-49-6
IUPAC Name:
3-fluoro-5-(trifluoromethoxy)benzoic acid
Synonyms:
3-fluoro-5-(trifluoromethoxy)benzoic acid
Molecular Weight (g/mol):
224.11
Molecular Formula:
C8H4F4O3
InChi Key:
RPEBABGCYLHTAK-UHFFFAOYSA-N
SMILES:
OC(=O)C1=CC(OC(F)(F)F)=CC(F)=C1
1
–
1
of
1
results
1
3-Fluoro-5-(trifluoromethoxy)benzoic acid, 97%, Thermo Scientific™
CAS: 1242258-49-6 Molecular Formula: C8H4F4O3 Molecular Weight (g/mol): 224.11 MDL Number: MFCD18783221 InChI Key: RPEBABGCYLHTAK-UHFFFAOYSA-N Synonym: 3-fluoro-5-trifluoromethoxy benzoic acid PubChem CID: 66523531 IUPAC Name: 3-fluoro-5-(trifluoromethoxy)benzoic acid SMILES: OC(=O)C1=CC(OC(F)(F)F)=CC(F)=C1