accessibility menu, dialog, popup

UserName

CAS RN 1238864-53-3

H3CNHNNH2HCl

CAS RN 1238864-53-3

IUPAC Name: 5-ethyl-1H-pyrazol-3-amine hydrochloride
Synonyms: 5-ethyl-1H-pyrazol-3-amine hydrochloride
Molecular Weight (g/mol): 147.61
Molecular Formula: C5H10ClN3
InChi Key: UXXRQTUGVMELDL-UHFFFAOYSA-N
SMILES: Cl.CCC1=CC(N)=NN1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results