CAS RN 123606-70-2
CAS RN 123606-70-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
3
of
3
results
1
3-(3-Azido-2,3-dideoxy-Beta-D-erythro-pentofuranosyl)-5-methyl-2,4(1H,3H)-pyrimidinedione, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
3-(3-Azido-2,3-dideoxy-Beta-D-erythro-pentofuranosyl)-5-methyl-2,4(1H,3H)-pyrimidinedione, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
3-(3-Azido-2,3-dideoxy-b-D-erythro-pentofuranosyl)-5-methyl-2,4(1H,3H)-pyrimidinedione, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.