CAS RN 123524-52-7
CAS RN 123524-52-7
IUPAC Name:
3-[1-(diphenylmethyl)azetidin-3-yl] 5-propan-2-yl 2-amino-6-methyl-4-(3-nitrophenyl)-1,4-dihydropyridine-3,5-dicarboxylate
Synonyms:
azelnidipine
Molecular Weight (g/mol):
582.66
Molecular Formula:
C33H34N4O6
InChi Key:
ZKFQEACEUNWPMT-UHFFFAOYNA-N
SMILES:
CC(C)OC(=O)C1=C(C)NC(N)=C(C1C1=CC=CC(=C1)[N+]([O-])=O)C(=O)OC1CN(C1)C(C1=CC=CC=C1)C1=CC=CC=C1
1
–
1
of
1
results
1
Thermo Scientific Chemicals Azelnidipine
CAS: 123524-52-7 Molecular Formula: C33H34N4O6