CAS RN 123334-00-9
CAS RN 123334-00-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Titanium(IV) oxide sulfate sulfuric acid hydrate
CAS: 123334-00-9 Molecular Formula: TiOSO4.xH2O + H2SO4 MDL Number: MFCD00011270 Synonym: Titanyl sulfate,Titanium(IV) oxysulfate-sulfuric acid