accessibility menu, dialog, popup

UserName

CAS RN 123333-88-0

OFFFOOFFFOCu

CAS RN 123333-88-0

IUPAC Name: bis(trifluoroacetic acid) copper hydrate
Synonyms: bis(trifluoroacetic acid) copper hydrate
Molecular Weight (g/mol): 309.61
Molecular Formula: C4H4CuF6O5
InChi Key: UXLSNKPEIUIWAD-UHFFFAOYSA-N
SMILES: O.[Cu].OC(=O)C(F)(F)F.OC(=O)C(F)(F)F
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results