accessibility menu, dialog, popup

UserName

CAS RN 122547-49-3

(S)N(R)S(R)OOOOHHH(R)OHNaMe

CAS RN 122547-49-3

IUPAC Name: bis(6-(1-hydroxyethyl)-7-oxo-3-(oxolan-2-yl)-4-thia-1-azabicyclo[3.2.0]hept-2-ene-2-carboxylic acid) pentahydrate disodium
Synonyms: bis(6-(1-hydroxyethyl)-7-oxo-3-(oxolan-2-yl)-4-thia-1-azabicyclo[3.2.0]hept-2-ene-2-carboxylic acid) pentahydrate disodium
Molecular Weight (g/mol): 706.68
Molecular Formula: C24H40N2Na2O15S2
InChi Key: SMFQLYMKANPICV-UHFFFAOYNA-N
SMILES: O.O.O.O.O.[Na].[Na].CC(O)C1C2SC(C3CCCO3)=C(N2C1=O)C(O)=O.CC(O)C1C2SC(C3CCCO3)=C(N2C1=O)C(O)=O
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results