accessibility menu, dialog, popup

UserName

CAS RN 122-25-8

IUPAC Name: 5-[(3-carboxy-4-hydroxyphenyl)methyl]-2-hydroxybenzoic acid
Synonyms: 5-[(3-carboxy-4-hydroxyphenyl)methyl]-2-hydroxybenzoic acid
Molecular Weight (g/mol): 288.26
Molecular Formula: C15H12O6
InChi Key: JWQFKVGACKJIAV-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC(CC2=CC=C(O)C(=C2)C(O)=O)=CC=C1O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results