accessibility menu, dialog, popup

UserName

ClOCl

CAS RN 122-01-0

IUPAC Name: 4-chlorobenzoyl chloride
Synonyms: 4-chlorobenzoyl chloride
Molecular Weight (g/mol): 175.01
Molecular Formula: C7H4Cl2O
InChi Key: RKIDDEGICSMIJA-UHFFFAOYSA-N
SMILES: ClC(=O)C1=CC=C(Cl)C=C1
Molecular Weight (g/mol) 
  • (2)
  • (8)
Quantity 
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (3)
  • (4)
Percent Purity 
  • (3)
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (3)
  • (3)
  • (2)
  • (8)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (4)
  • (3)
  • (4)
  • (4)