CAS RN 1219805-73-8
CAS RN 1219805-73-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
Metaldehyde-d16, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
Metaldehyde-d16, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.