accessibility menu, dialog, popup

UserName

CH3OOCH3H3CCH3OOH3C

CAS RN 121823-85-6

IUPAC Name: 1,3-diethyl 2-methyl-2-(3-methylbutyl)propanedioate
Synonyms: 1,3-diethyl 2-methyl-2-(3-methylbutyl)propanedioate
Molecular Weight (g/mol): 244.33
Molecular Formula: C13H24O4
InChi Key: LFMBZEGLMSBWHG-UHFFFAOYSA-N
SMILES: CCOC(=O)C(C)(CCC(C)C)C(=O)OCC
Grade 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)