accessibility menu, dialog, popup

UserName

CH3NSN

CAS RN 1217863-27-8

IUPAC Name: 2-(5-methyl-4-phenyl-1,3-thiazol-2-yl)pyridine
Synonyms: 2-(5-methyl-4-phenyl-1,3-thiazol-2-yl)pyridine
Molecular Weight (g/mol): 252.34
Molecular Formula: C15H12N2S
InChi Key: ZTYJKDZBAFCCRT-UHFFFAOYSA-N
SMILES: CC1=C(N=C(S1)C1=CC=CC=N1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results