CAS RN 1216745-60-6
CAS RN 1216745-60-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
rac Methotrimeprazine-d3 Hydrochloride, TRC
CAS: 1216745-60-6 Molecular Formula: C19H22D3ClN2OS Molecular Weight (g/mol): 367.95 Synonym: 2-Methoxy-N,N,β-trimethyl-10H-phenothiazine-10-propanamine-d3 Hydrochloride,10-[3-(Dimethylamino)-2-methylpropyl]-2-methoxyphenothiazine-d3 MonoHydrochloride,2-Methoxytrimeprazine-d3 Hydrochloride,2-Methoxytrimeprazine-d3 SMILES: CN(C)CC(C)CN1C2=C(C=CC(OC([2H])([2H])[2H])=C2)SC3=CC=CC=C31.Cl