accessibility menu, dialog, popup

UserName

POOOOPOOOOPOOOOOHCaCaCaCaCa

CAS RN 12167-74-7

IUPAC Name: tricalcium diphosphate
Synonyms: calcium phosphate tricalcium phosphate tricalcium diphosphate calcium orthophosphate tricalcium bis(phosphate) tricalcium orthophosphate tertiary calcium phosphate tribasic calcium phosphate calcigenol simple calcium phosphate tribasic
Molecular Weight (g/mol): 310.17
Molecular Formula: Ca3O8P2
InChi Key: QORWJWZARLRLPR-UHFFFAOYSA-H
SMILES: [Ca++].[Ca++].[Ca++].[O-]P([O-])([O-])=O.[O-]P([O-])([O-])=O
Molecular Weight (g/mol) 
  • (6)
  • (3)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (1)
  • (6)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (3)