accessibility menu, dialog, popup

UserName

CAS RN 1215206-32-8

IUPAC Name: 3'-bromo-4'-methoxy-[1,1'-biphenyl]-3-carboxylic acid
Synonyms: 3'-bromo-4'-methoxy-[1,1'-biphenyl]-3-carboxylic acid
Molecular Weight (g/mol): 307.14
Molecular Formula: C14H11BrO3
InChi Key: AAYOTNRWOUSBAC-UHFFFAOYSA-N
SMILES: COC1=C(Br)C=C(C=C1)C1=CC(=CC=C1)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results